Phenyl phosphorodichloridate, 97%
Phenyl phosphorodichloridate, 97%
Phenyl phosphorodichloridate, 97%
Thermo Scientific Chemicals

Phenyl phosphorodichloridate, 97%

CAS: 770-12-7 | C6H5Cl2O2P | 210.978 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10479.22
または、製品番号A10479-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS770-12-7
IUPAC Namephenyl chlorophosphonochloridate
Molecular FormulaC6H5Cl2O2P
InChI KeyTXFOLHZMICYNRM-UHFFFAOYSA-N
SMILESClP(Cl)(=O)OC1=CC=CC=C1
さらに表示
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5195-1.5245 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥96.0%
Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent for preparation of phosphate diesters.Phenyl phosphorodichloridate is used in the preparation of symmetrical phosphate diesters. It is used as a phosphorylating agent for alcohols and amines. It is also involved in the preparation of phenylphosphoro di-(1-imidazolidate) and 2-phenyl-bis-triazoloylphosphate, which is used in peptide synthesis. It plays a vital role in the Pfitzner-Moffatt oxidation which involves oxidation of alcohols to the corresponding aldehydes and ketones. Furthermore, it is utilized in Beckmann rearrangement for the preparation of N-phenylacetamide from acetophenone oxime.

Solubility
Immiscible with water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, alcohols and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Reagent for the preparation of phosphate diesters. The phenyl group is removable by hydrogenolysis: J. Am. Chem. Soc., 75, 4510 (1953).
  2. Coupling reagent, e.g. in pyridine for the preparation of derivatives of sulfinic acids with alcohols, amines, thiols etc.: Synthesis, 937 (1980). Similarly, carboxylic acids in pyridine can be coupled with thiols to give thiocarboxylic acid S-esters: Can. J. Chem., 58, 2645 (1980), and with sodium azide in the presence of a phase-transfer catalyst to give acyl azides: Synth. Commun., 13, 289 (1983).
  3. Reacts with the Li enolates of ß-diketones, in the presence of LiCl, to give ß-chloro-ɑß-enones: Can. J. Chem., 64, 520 (1986).
  4. In combination with NaI, cleaves dialkyl ethers to iodides in high yield: Synth. Commun., 18, 119 (1988), and 1,3-dithianes and other dithioacetals to carbonyl groups: Tetrahedron Lett., 29, 5471 (1988).
  5. In combination with Dimethyl sulfoxide, A13280, effects selective oxidation of alcohols to aldehydes or ketones with less ɑ-chlorination than the more usual Swern (Oxalyl chloride, A18012) system: J. Org. Chem., 52, 5621 (1987). Similarly, benzylamines have been converted to benzaldehydes: Synth. Commun., 19, 3407 (1989).
  6. For a brief feature on uses of the reagent in synthesis, see: Synlett, 1651 (2004).
  7. Kuo, C. W.; Hsieh, M. T.; Gao, S.; Shao, Y. M.; Yao, C. F.; Shia, K. S. Beckmann Rearrangement of Ketoximes Induced by Phenyl Dichlorophosphate at Ambient Temperature. Molecules 2012, 17 (11), 13662-13672.
  8. Nagarkar, A. A.; Crochet, A.; Fromm, K. M.; Kilbinger, A. F. Efficient amine end-functionalization of living ring-opening metathesis polymers. Macromolecules 2012, 45 (11), 4447-4453.