Benzamide, 98+%
Benzamide, 98+%
Benzamide, 98+%
Thermo Scientific Chemicals

Benzamide, 98+%

CAS: 55-21-0 | C7H7NO | 121.139 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A10501.22
または、製品番号A10501-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS55-21-0
IUPAC Namebenzamide
Molecular FormulaC7H7NO
InChI KeyKXDAEFPNCMNJSK-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Melting Point (clear melt)125-129°C
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (GC)≥98.0%
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzamide is utilized to study the mechanism of photocatalytic decomposition of aqueous solution of acetic acid, acetamide and acetonitrile in the presence of semiconductors. It is used as a nictoinamide-mimic PARP inhibitor and neuroprotectant. Further, it is used to develop a robust screening method to study biotransformations using (+)-gamma-lactamase enzyme. It is also employed in the determination of glycine. In addition to this, it is used as an intermediate in organic synthesis as well as in the production of pharmaceuticals and dyes.

Solubility
Soluble in water, methanol, ammonia, pyridine, carbon tetrachloride, carbon disulfide and ethanol. Slightly soluble in benzene and ethyl ether.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

General References:

  1. Reduction with lithium aluminum hydride gives benzylamine. By using lithium tris(diethylamino)aluminum hydride (see Diethyl amine, A11716), selective reduction to benzaldehyde is possible: Tetrahedron Lett., 32, 6903 (1991).
  2. An exceptionally mild method for dehydration of amides to nitriles by transamidation with acetonitrile in the presence of an aldehyde and formic acid is exemplified by the high yield conversion of benzamide to benzonitrile: J. Org. Chem., 61, 6486 (1996).
  3. Barrett, K. T.; Miller, S. J. Regioselective Derivatizations of a Tribrominated Atropisomeric Benzamide Scaffold. Org. Lett. 2015, 17 (3), 580-583.
  4. Manna, S.; Antonchick, A. P. Organocatalytic Oxidative Annulation of Benzamide Derivatives with Alkynes. Angew. Chem. Int. Ed. 2014, 53 (28), 7324-7327.