Valerophenone, 99%
Valerophenone, 99%
Valerophenone, 99%
Thermo Scientific Chemicals

Valerophenone, 99%

CAS: 1009-14-9 | C11H14O | 162.23 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10525.22
または、製品番号A10525-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1009-14-9
IUPAC Name1-phenylpentan-1-one
Molecular FormulaC11H14O
InChI KeyXKGLSKVNOSHTAD-UHFFFAOYSA-N
SMILESCCCCC(=O)C1=CC=CC=C1
さらに表示
Appearance (Color)Clear, colourless to pale yellow
Assay (GC)>98.5%
Formliquid
Refractive Index1.5130-1.5160
Valerophenone is used as intermediates of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Valerophenone is used as intermediates of liquid crystals.

Solubility
Insoluble in water.

Notes
 Incompatible with strong oxidizing agents, acids, bases, plastics. Store at room temperature with proper ventilation.
RUO – Research Use Only

General References:

  1. R.G.Zepp; M.M.Gumz; W.L.Miller; H.Gao. Photoreaction of Valerophenone in Aqueous Solution. J. Phys. Chem. A. 1998, 102, (28),5716-5723
  2. P.Novak; J.Matousek; J.Briza. Valerophenone synthase-like chalcone synthase homologues in Humulus lupulus. Biologia Plantarum. 2003, 46, (3),375-381