Dimethyl aminoterephthalate, 99%
Dimethyl aminoterephthalate, 99%
Dimethyl aminoterephthalate, 99%
Dimethyl aminoterephthalate, 99%
Thermo Scientific Chemicals

Dimethyl aminoterephthalate, 99%

CAS: 5372-81-6 | C10H11NO4 | 209.201 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A10567.0B
または、製品番号A10567-0B
価格(JPY)
-
見積もりを依頼する
数量:
1000 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5372-81-6
IUPAC Name1,4-dimethyl 2-aminobenzene-1,4-dicarboxylate
Molecular FormulaC10H11NO4
InChI KeyDSSKDXUDARIMTR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C(=O)OC)C(N)=C1
さらに表示
Assay (GC)≥98.5%
Appearance (Color)Cream to yellow to green to brown
FormPowder
Melting Point (clear melt)129.0-137.0?C
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl aminoterephthalate is used in the preparation of imidazotetrazinones as probes for the action of temozolomide. It is also used to prepare aminoquinazolines with neurokinin-2 receptor antagonist activity.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Borbone, F.; Tarallo, O.; Roviello, V.; Caruso, U.; Pirozzi, B.; Roviello, A. Novel rigid rod polymers from a thermal cyclization reaction. Eur. Polym. J. 2015, 63, 80-89.
  2. Samokhvalov, A. Adsorption on Mesoporous Metal-Organic Frameworks in Solution: Aromatic and Heterocyclic Compounds. Chem. Eur. J. 2015, 21 (47), 16726-16742.