Tetraphenylphosphonium chloride, 98%
Tetraphenylphosphonium chloride, 98%
Tetraphenylphosphonium chloride, 98%
Thermo Scientific Chemicals

Tetraphenylphosphonium chloride, 98%

CAS: 2001-45-8 | C24H20ClP | 374.85 g/mol
製品番号(カタログ番号) A10575.09
または、製品番号A10575-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2001-45-8
IUPAC Nametetraphenylphosphanium chloride
Molecular FormulaC24H20ClP
InChI KeyWAGFXJQAIZNSEQ-UHFFFAOYSA-M
SMILES[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)White to cream
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)268-287?C
Tetraphenylphosphonium chloride is used to generate lipophilic salts from inorganic and organometallic anions. Thus, Ph4P+ is useful as a phase-transfer catalyst, again because it allows inorganic anions to dissolve in organic solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraphenylphosphonium chloride is used to generate lipophilic salts from inorganic and organometallic anions. Thus, Ph4P+ is useful as a phase-transfer catalyst, again because it allows inorganic anions to dissolve in organic solvents.

Solubility
Soluble in water.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. David R.Wood; Fred W.Reimherr; Paul H.Wender. Mechanism of inhibition of fadu hypopharyngeal carcinoma cell growth by tetraphenylphosphonium chloride. International Journal of Cancer. 1994, 57, (2),247-253
  2. J.T.van Elteren; N.G.Haselager; H.A.Das C.L. de Ligny; J.Agterdenbos. Determination of arsenate in aqueous samples by precipitation of the arsenic(V)—molybdate complex with tetraphenylphosphonium chloride and neutron activation analysis or hydride generation atomic absorption spectrometry. Analytica Chimica Acta. 1991, 252, (1-2),89-95
  3. Reagent for Bi and Co.
  4. High-temperature phase-transfer catalyst (see Appendix 2). In combination with Bis(acetonitrile) dichloropalladium(II) , 10002, provides an effective catalyst for the Heck reaction of normally unreactive aryl halides, e.g. chlorobenzene with styrene to give stilbene. The reaction is performed at 140-150° in DMF or NMP in the presence of sodium acetate. The reaction may be further improved by the addition of 10-12 mol% N,N-Dimethyl glycine, A19670: Angew. Chem. Int. Ed., 37, 481 (1998).