1-Bromononane, 99%
1-Bromononane, 99%
1-Bromononane, 99%
Thermo Scientific Chemicals

1-Bromononane, 99%

CAS: 693-58-3 | C9H19Br | 207.155 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10577.14
または、製品番号A10577-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS693-58-3
IUPAC Name1-bromononane
Molecular FormulaC9H19Br
InChI KeyAYMUQTNXKPEMLM-UHFFFAOYSA-N
SMILESCCCCCCCCCBr
さらに表示
Assay (GC)≥98.5%
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4530-1.4560 @ 20?C
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromononane is used as organic intermediates. It is mainly used in pharmaceutical synthesis and industrial dye intermediates.

Solubility
Insoluble in water. Soluble in ethyl ether and benzene.

Notes
Stable under normal conditions. Incompatible with oxidizing agents.
RUO – Research Use Only