2,4-Dimethylbenzoic acid, 97%
2,4-Dimethylbenzoic acid, 97%
2,4-Dimethylbenzoic acid, 97%
Thermo Scientific Chemicals

2,4-Dimethylbenzoic acid, 97%

CAS: 611-01-8 | C9H10O2 | 150.18 g/mol
製品番号(カタログ番号) A10589.09
または、製品番号A10589-09
価格(JPY)
-
数量:
10 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS611-01-8
IUPAC Name2,4-dimethylbenzoic acid
Molecular FormulaC9H10O2
InChI KeyBKYWPNROPGQIFZ-UHFFFAOYSA-N
SMILESCC1=CC=C(C(O)=O)C(C)=C1
さらに表示
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White to pale cream
Assay (Silylated GC)≥97.5%
Melting Point (clear melt)120.0-126.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only