5-Nitroindole, 99%
5-Nitroindole, 99%
5-Nitroindole, 99%
Thermo Scientific Chemicals

5-Nitroindole, 99%

CAS: 6146-52-7 | C8H6N2O2 | 162.148 g/mol
製品番号(カタログ番号) A10608.14
または、製品番号A10608-14
価格(JPY)
-
見積もりを依頼する
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6146-52-7
IUPAC Name5-nitro-1H-indole
Molecular FormulaC8H6N2O2
InChI KeyOZFPSOBLQZPIAV-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2NC=CC2=C1
さらに表示
Appearance (Color)Yellow
FormCrystals or powder or crystalline powder
Melting Point (clear melt)137.0-144.0?C
Assay (GC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only