5-Methyl-1H-benzotriazole, 98+%
5-Methyl-1H-benzotriazole, 98+%
5-Methyl-1H-benzotriazole, 98+%
Thermo Scientific Chemicals

5-Methyl-1H-benzotriazole, 98+%

CAS: 136-85-6 | C7H7N3 | 133.15 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10615.22
または、製品番号A10615-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS136-85-6
Assay (GC)≥98.0%
Appearance (Color)White to pale cream to pale brown
FormPowder
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Devon A.Cancilla; J.Christopher Baird, Steven W.Geis; Steven R.Corsi. Studies of the environmental fate and effect of aircraft deicing fluids: Detection of 5-methyl-1H-benzotriazole in the fathead minnow (Pimephales promelas). Environmental Toxicology and Chemistry. 2003, 22, (1),134-140
  2. Alexander Muller; Stefan C.Weiss; Judith Beißwenger; H.Georg Leukhardt; Wolfgang Schulz ; Wolfram Seitz; Wolfgang K.L.Ruck, Walter H.Weber. Identification of ozonation by-products of 4- and 5-methyl-1H-benzotriazole during the treatment of surface water to drinking water . Water Research. 2012, 46, (3),679-690