Thermo Scientific Chemicals

Acetaldoxime, syn + anti, 98%, Thermo Scientific Chemicals

Catalog number: A10640.30
250 g, Each
Thermo Scientific Chemicals

Acetaldoxime, syn + anti, 98%, Thermo Scientific Chemicals

Catalog number: A10640.30
250 g, Each
Quantity
Catalog number: A10640.30
also known as A10640-30
Price (USD)
Price: 438.00
Online price: 378.65
Your price:
Quantity
-

Chemical Identifiers

CAS
107-29-9
IUPAC Name
(E)-N-ethylidenehydroxylamine
Molecular Formula
C2H5NO
InChI Key
FZENGILVLUJGJX-NSCUHMNNSA-N
SMILES
C\C=N\O
Form
liquid
Comment
Physical state varies with syn/anti ratio
Appearance (Color)
White solid or clear, colourless
Assay (GC)
> 98.0% (sum of isomers)

Description

Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetaldoxime is used as an oxygen scavenger in boiler water. It is also used as an intermediate in chemical synthesis and pharmaceuticals. It is involved in the rearrangement reaction to prepare acetamide by using nickel(II) acetate as a catalyst. It acts as a precursor to prepare heterocyclic compound such as spiroisoxazoline. Further, it is used to prepare alkylated(Z)-oximes by deprotonation followed by reaction with benzyl bromide.

Solubility
Soluble in water, ethanol and ether.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text