Thermo Scientific Chemicals

Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog number: A10645.18
50 g, Each
Thermo Scientific Chemicals

Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals

Catalog number: A10645.18
50 g, Each
Quantity
Catalog number: A10645.18
also known as A10645-18
Price (USD)
Price: 114.00
Online price: 98.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1080-32-6
IUPAC Name
diethyl benzylphosphonate
Molecular Formula
C11H17O3P
InChI Key
AIPRAPZUGUTQKX-UHFFFAOYSA-N
SMILES
CCOP(=O)(CC1=CC=CC=C1)OCC
Assay (GC)
≥98.5%
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4950-1.4980 @ 20?C
Form
Liquid

Description

Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl benzylphosphonate is used as a reactant for synthesis of 3,5-dihydroxy-4-isopropylstilbene for treatment of skin disorders and natural cytotoxic marine products of polyketide origin via intramolecular Diels-Alder reactions. It is also used as reactant for cyclization of aryl ethers, amines and amides & for investigating the effects of functional groups on the performance of clue organic LEDs.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text