2-Chloroacetophenone, 98%, Thermo Scientific Chemicals
2-Chloroacetophenone, 98%, Thermo Scientific Chemicals
2-Chloroacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloroacetophenone, 98%, Thermo Scientific Chemicals

製品番号(カタログ番号) A10749.22
または、製品番号A10749-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS532-27-4
IUPAC Name2-chloro-1-phenylethan-1-one
Molecular FormulaC8H7ClO
InChI KeyIMACFCSSMIZSPP-UHFFFAOYSA-N
SMILESClCC(=O)C1=CC=CC=C1
さらに表示
Solution TestClear to slightly turbid colorless to yellow (10% in Ethanol, non-U.S. specification)
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (GC)≥97.5%
Melting Point (clear melt)51.5-57.5°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only