Agar powder
Agar powder
Agar powder
Thermo Scientific Chemicals

Agar powder

CAS: 9002-18-0 | C14H24O9 | 336.337 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A10752.0E
または、製品番号A10752-0E
価格(JPY)
-
見積もりを依頼する
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9002-18-0
IUPAC Name2-({4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl}oxy)-6-(hydroxymethyl)-4-methoxyoxane-3,5-diol
Molecular FormulaC14H24O9
InChI KeyGYYDPBCUIJTIBM-UHFFFAOYNA-N
SMILESCOC1C(O)C(CO)OC(OC2C3COC2C(O)C(C)O3)C1O
さらに表示
Appearance (Color)White to cream or pale yellow to pale brown
FormPowder
CommentSpecification differs for U.S. and non-U.S. material where indicated
Gel strength≥500 g/cm² (1.5 % @ 20°C)
Residual water≤20%
さらに表示
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.

Solubility
Soluble in water at temperatures between 950C and 1000 C. Insoluble in cold water.

Notes
Prolonged exposure to high temperatures can degrade Agar powder.
RUO – Research Use Only

General References:

  1. For gel preparation.
  2. Chi, W. J.; Chang, Y. K.; Hong, S. K. Agar degradation by microorganisms and agar-degrading enzymes. Appl. Microbiol. Biotechnol. 2012, 94 (4), 917-30.
  3. Laurienzo, P. Marine polysaccharides in pharmaceutical applications: An overview. Mar. Drugs. 2010, 8 (9), 2435-2465.