(1-Dodecyl)trimethylammonium bromide, 99%
(1-Dodecyl)trimethylammonium bromide, 99%
(1-Dodecyl)trimethylammonium bromide, 99%
Thermo Scientific Chemicals

(1-Dodecyl)trimethylammonium bromide, 99%

CAS: 1119-94-4 | C15H34BrN | 308.348 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A10761.30
または、製品番号A10761-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1119-94-4
IUPAC Namedodecyltrimethylazanium bromide
Molecular FormulaC15H34BrN
InChI KeyXJWSAJYUBXQQDR-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCC[N+](C)(C)C
さらに表示
Appearance (Color)White to cream
Assay (Titration ex Bromide)>98.5 to <101.5%
FormPowder
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Dodecyl)trimethylammonium bromide is used as a surfactant, as an emulsifier of rubber and asphalt and as an anti-static agent of synthetic fiber. It acts as an ionic surfactant and useful in the preparation of gold nanopartilces along with sodium dodecylsulfate. It is also used as paint strippers and foaming stabilizers. It is used to prepare dodecyltrimethylammonium bromide. By using conductimetry, density measurements, and small angle neutron scattering, it is used to study the micelle formation in water-dimethylsulfoxide mixture. Further, it is a useful biochemical for proteomics research.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Wang, P. Y.; Shah, I.; Huang, C. P. Preparation and characterization of functionalized poly(vinyl chloride) membranes for selective separation of perchlorate from water. J. Membr. Sci. 2015, 476, 561-570.
  2. Al-rashdi, A. A. A submicron mesoporous silica for the determination of organosulphur in sea water. Anal. Chem. Res. 2015, 4, 25-32.