o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals
o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals
o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

o-Toluoyl chloride, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号)数量
A10775.22
または、製品番号A10775-22
100 g
製品番号(カタログ番号) A10775.22
または、製品番号A10775-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS933-88-0
IUPAC Name2-methylbenzoyl chloride
Molecular FormulaC8H7ClO
InChI KeyGPZXFICWCMCQPF-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(Cl)=O
さらに表示
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.5520-1.5560 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
o-Toluoyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluoyl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store under dry inert gas. Store in cool, dry conditions in well sealed containers. Incompatible with bases, active metals, alcohols,amines and oxidizing agents. It is sensitive to moisture.
RUO – Research Use Only

General References:

  1. P. S. Hofman.; D. J. Reiding.; W. Th. Nauta. Synthesis of ortho-substituted benzophenones by way of benzophenones carrying A t-butyl group as a blocking group. Recueil des Travaux Chimiques des Pays-Bas. 1960, 79, (8), 790-793.
  2. S. Icli.; H. Icil.; S. Alpb, H. Koc.; A. Mckillop. NMR, Absorption and Fluorescence Parameters of Azlactones. Spectroscopy Letters: An International Journal for Rapid Communication. 1994, 27 (9), 1115-1128.