Thermo Scientific Chemicals

Acetone oxime, 98%, Thermo Scientific Chemicals

Catalog number: A10802.0B
1000 g, Each
Thermo Scientific Chemicals

Acetone oxime, 98%, Thermo Scientific Chemicals

Catalog number: A10802.0B
1000 g, Each
Quantity
Catalog number: A10802.0B
also known as A10802-0B
Price (USD)
Price: 282.00
Online price: 243.65
Your price:
Quantity
-

Chemical Identifiers

CAS
127-06-0
IUPAC Name
N-(propan-2-ylidene)hydroxylamine
Molecular Formula
C3H7NO
InChI Key
PXAJQJMDEXJWFB-UHFFFAOYSA-N
SMILES
CC(C)=NO
Assay (GC)
≥97.5%
Appearance (Color)
White to cream
Melting Point (clear melt)
57.5-63.5?C
Form
Crystals or powder or crystalline powder or fused solid

Description

It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

Solubility
Soluble in water (330 g/L at 20°C).

Notes
Store away from oxidizing agents. Protect against electrostatic charges.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text