2-Aminobenzhydrazide, 98+%
2-Aminobenzhydrazide, 98+%
2-Aminobenzhydrazide, 98+%
Thermo Scientific Chemicals

2-Aminobenzhydrazide, 98+%

CAS: 1904-58-1 | C7H9N3O | 151.17 g/mol
製品番号(カタログ番号) A10805.18
または、製品番号A10805-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1904-58-1
IUPAC Name2-aminobenzohydrazide
Molecular FormulaC7H9N3O
InChI KeyGRWMSCBKWMQPON-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=CC=C1N
さらに表示
Assay (GC)>98.0%
FormCrystals or crystalline powder
Appearance (Color)White to cream to pink to brown
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only