4-Chloro-o-phenylenediamine, 97%
4-Chloro-o-phenylenediamine, 97%
4-Chloro-o-phenylenediamine, 97%
4-Chloro-o-phenylenediamine, 97%
Thermo Scientific Chemicals

4-Chloro-o-phenylenediamine, 97%

CAS: 95-83-0 | C6H7ClN2 | 142.59 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A10823.36
または、製品番号A10823-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-83-0
IUPAC Name4-chlorobenzene-1,2-diamine
Molecular FormulaC6H7ClN2
InChI KeyBXIXXXYDDJVHDL-UHFFFAOYSA-N
SMILESNC1=CC=C(Cl)C=C1N
さらに表示
FormPowder
Melting Point (clear melt)68.0-77.0?C
Appearance (Color)Pale brown to brown
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only