3-Acetylbenzonitrile, 97+%
3-Acetylbenzonitrile, 97+%
3-Acetylbenzonitrile, 97+%
Thermo Scientific Chemicals

3-Acetylbenzonitrile, 97+%

CAS: 6136-68-1 | C9H7NO | 145.16 g/mol
製品番号(カタログ番号) A10841.06
または、製品番号A10841-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS6136-68-1
Appearance (Color)White to cream or pale yellow to pale brown
FormPowder
Assay (GC)≥97.0%
Melting Point (clear melt)94.0-103.0°C
Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Preparation of (q2-2-Acetyl-4-cyanophenyl)tetra- carbonylmanganese by Reaction of 3-Acetylbenzonitrile with Benzylpentacarbonylmanganese. Four aromatic amidoximes were synthesized by reacting hydroxylamine with 1,3 -dicyanobenzene, 1,4- dicyanobenzene, 3-acetylbenzonitrile, and 4-acetylbenzonitrile. Darzens condensation of 3-acetylbenzonitrile provided aldehyde.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Luísa M.P.F. Amaral,; Piotr Szterner,; Victor M.F. Morais,; Manuel A.V. Ribeiro da Silva. Thermochemical study of the isomeric compounds: 3-acetylbenzonitrile and benzoylacetonitrile. The Journal of Chemical Thermodynamics 2015, 91452-458.
  2. Corrine Y. Watsona,; William J.D. Whishb,; Michael D. Threadgilla. Synthesis of 3-Substituted Benzamides and 5-Substituted Isoquinolin-1(2H)-ones and Preliminary Evaluation as Inhibitors of Poly(ADP-ribose)polymerase (PARP) . Bioorganic & Medicinal Chemistry. 1998, 6 (6), 721-734.