Thermo Scientific Chemicals

4-Bromo-N,N-dimethylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A10854.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-N,N-dimethylaniline, 98+%, Thermo Scientific Chemicals

Catalog number: A10854.14
25 g, Each
Quantity
Catalog number: A10854.14
also known as A10854-14
Price (USD)
Quantity
-

Chemical Identifiers

CAS
586-77-6
IUPAC Name
4-bromo-N,N-dimethylaniline
Molecular Formula
C8H10BrN
InChI Key
XYZWMVYYUIMRIZ-UHFFFAOYSA-N
SMILES
CN(C)C1=CC=C(Br)C=C1
Appearance (Color)
White to pale brown or blue to gray
Melting Point (clear melt)
50.0-57.0?C
Assay (GC)
≥98.0%
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder or fused solid

Description

4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-N,N-dimethylaniline is used as an internal standard in the determination of iodine present as iodide (as in pharmaceuticals), iodate (as in iodized table salt) and covalently bound to organic compounds (as in milk and vegetables).

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, acids and metals. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text