Isonicotinoyl chloride hydrochloride, 97%
Isonicotinoyl chloride hydrochloride, 97%
Isonicotinoyl chloride hydrochloride, 97%
Thermo Scientific Chemicals

Isonicotinoyl chloride hydrochloride, 97%

CAS: 39178-35-3 | C6H5Cl2NO | 178.01 g/mol
数量:
10 g
50 g
250 g
製品番号(カタログ番号) A10863.30
または、製品番号A10863-30
価格(JPY)
-
数量:
250 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS39178-35-3
IUPAC Namehydrogen pyridine-4-carbonyl chloride chloride
Molecular FormulaC6H5Cl2NO
InChI KeyBNTRVUUJBGBGLZ-UHFFFAOYSA-N
SMILES[H+].[Cl-].ClC(=O)C1=CC=NC=C1
さらに表示
Appearance (Color)White to cream to pale brown or pale yellow
FormCrystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)153-163°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only