2,3-Dimethylaniline, 98%
2,3-Dimethylaniline, 98%
2,3-Dimethylaniline, 98%
Thermo Scientific Chemicals

2,3-Dimethylaniline, 98%

CAS: 87-59-2 | C8H11N | 121.183 g/mol
製品番号(カタログ番号) A10918.36
または、製品番号A10918-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS87-59-2
IUPAC Name2,3-dimethylaniline
Molecular FormulaC8H11N
InChI KeyVVAKEQGKZNKUSU-UHFFFAOYSA-N
SMILESCC1=CC=CC(N)=C1C
さらに表示
Appearance (Color)Clear colorless to yellow or orange or red-brown or yellow-brown
FormLiquid
Identification (FTIR)Conforms
Assay (GC)≥97.5%
Refractive Index1.5660-1.5700 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only