1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%
Thermo Scientific Chemicals

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide, 98%

CAS: 22572-40-3 | C9H20N3+ | 170.28 g/mol
製品番号(カタログ番号) A10962.14
または、製品番号A10962-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS22572-40-3
IUPAC Name(3-{[(ethylimino)methylidene]amino}propyl)trimethylazanium iodide
Molecular FormulaC9H20IN3
InChI KeyAGSKWMRPXWHSPF-UHFFFAOYSA-M
SMILES[I-].CCN=C=NCCC[N+](C)(C)C
さらに表示
FormCrystalline powder or powder
Assay (Titration ex Iodide)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Appearance (Color)White to cream to yellow
Water Content (Karl Fischer Titration)≤1.0%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide methiodide is in proteins as water soluble carboxyl modifying reagent. It is used as a crosslinking agent.

Solubility
Soluble in water.

Notes
Keep in a dry, cool and well-ventilated place. Keep container tightly closed. Incompatible with acids, strong oxidizing agents, moist air and water.
RUO – Research Use Only

General References:

  1. Peptide coupling reagent: both the reagent and the urea by-product are soluble in neutral aqueous media.
  2. Jiang, J.; Yang, E.; Reddy, K. R.; Niedzwiedzki, D. M.; Kirmaier, C.; Bocian, D. F.; Holten, D.; Lindsey, J. S. Synthetic bacteriochlorins bearing polar motifs (carboxylate, phosphonate, ammonium and a short PEG). Water-solubilization, bioconjugation, and photophysical properties. New J. Chem. 2015, 39, 5694-5714.
  3. Ren, T.; Liu, Q.; Lu, H.; Liu, H.; Zhang, X.; Du, J. Multifunctional polymer vesicles for ultrasensitive magnetic resonance imaging and drug delivery. J. Mater. Chem. 2012, 22, 12329-12338.