Pyrazine-2-carbonitrile, 97+%
Pyrazine-2-carbonitrile, 97+%
Pyrazine-2-carbonitrile, 97+%
Thermo Scientific Chemicals

Pyrazine-2-carbonitrile, 97+%

CAS: 19847-12-2 | C5H3N3 | 105.10 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A10974.14
または、製品番号A10974-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS19847-12-2
IUPAC Namepyrazine-2-carbonitrile
Molecular FormulaC5H3N3
InChI KeyPMSVVUSIPKHUMT-UHFFFAOYSA-N
SMILESN#CC1=CN=CC=N1
さらに表示
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.0%
Refractive Index1.5325-1.5375 @ 20?C
Identification (FTIR)Conforms
FormLiquid
Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrazine-2-carbonitrile is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Tetsuya Sunahara; Satoru Onaka; Mitsuhiro Ito; Hiroyuki Imai; Katsuya Inoue; Tomoji Ozeki. Construction of nano-channels based on zinc(II) pyrazine-2-carboxylate complexes . European Journal of Inorganic Chemistry. 2004, 2004,(24), 4882-4890.
  2. Ying Tao; Jian-Rong Li; Qun Yu; Wei-Chao Song; Xiao-Lan Tong; Xian-He Bu. Coordination architectures of 2-(1H-tetrazol-5-yl)pyrazine with group IIB metal ions: luminescence and structural dependence on the metal ions and preparing conditions. CrystEngComm. 2008, 10,(6), 699-705.