Thermo Scientific Chemicals

Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech., Thermo Scientific Chemicals

Catalog number: A10982.36
500 g, Each
Thermo Scientific Chemicals

Methyl-5-norbornene-2,3-dicarboxylic anhydride, mixture of isomers, tech., Thermo Scientific Chemicals

Catalog number: A10982.36
500 g, Each
Quantity
Catalog number: A10982.36
also known as A10982-36
Price (USD)
Price: 102.00
Online price: 86.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25134-21-8
Appearance (Color)
Clear, colourless to pale yellow
Refractive Index
1.5005 - 1.5095 @20?C
Form
liquid
Assay (GC)
> 88.0% (sum of isomers)

Description

Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-5-norbornene-2,3-dicarboxylic anhydride is used as a curing agent in epoxy resins and in the preparation of vertically aligned and penetrated carbon nanotube/polymer film. It serves as an intermediate in polyesters, alkyd resins and plasticizers. It is also used in electrical laminating and filament-winding. Further, it is used in the preparation of methyl nadimides endcapped resins based on tris(3-aminophenyl)phosphine oxide. It is utilized in the flame retarding treatment.

Solubility
Miscible with acetone, benzene, naphtha and xylene.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text