4'-Ethoxyacetophenone, 98%
4'-Ethoxyacetophenone, 98%
4'-Ethoxyacetophenone, 98%
Thermo Scientific Chemicals

4'-Ethoxyacetophenone, 98%

CAS: 1676-63-7 | C10H12O2 | 164.20 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A11031.14
または、製品番号A11031-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1676-63-7
IUPAC Name1-(4-ethoxyphenyl)ethan-1-one
Molecular FormulaC10H12O2
InChI KeyYJFNFQHMQJCPRG-UHFFFAOYSA-N
SMILESCCOC1=CC=C(C=C1)C(C)=O
さらに表示
Appearance (Color)White to yellow
FormCrystalline powder, crystals and/or chunks
Assay (GC)≥97.5%
Melting Point (clear melt)34-38?C
4'-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Ethoxyacetophenone is used to produce 4-(4-ethoxy-phenyl)-thiazol-2-ylamine.

Solubility
Soluble in alcohol, water(791.1 mg/L).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Malav Kapadia,; Mihir Patel,; Jayantilal Joshi. Thermal, Catalytic and Antimicrobial Aspects of Poly[(2-hydroxy,4-ethoxyacetophenone)butylene] Resin and its Polychelates with 4f - block Elements. High Performance Polymers. 2008,