Thermo Scientific Chemicals

3-Iodobenzyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: A11140.14
25 g, Each
Thermo Scientific Chemicals

3-Iodobenzyl bromide, 96%, Thermo Scientific Chemicals

Catalog number: A11140.14
25 g, Each
Quantity
Catalog number: A11140.14
also known as A11140-14
Price (USD)
Price: 328.00
Online price: 280.65
Your price:
Quantity
-

Chemical Identifiers

CAS
49617-83-6
IUPAC Name
1-(bromomethyl)-3-iodobenzene
Molecular Formula
C7H6BrI
InChI Key
BACZSVQZBSCWIG-UHFFFAOYSA-N
SMILES
BrCC1=CC(I)=CC=C1
Appearance (Color)
White to cream or pale yellow to pale brown
Assay (GC)
≥95.0%
Form
Crystalline powder or powder
Melting Point (clear melt)
48-52°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text