Thermo Scientific Chemicals

Octanoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11149.AC
25 mL, Each
Thermo Scientific Chemicals

Octanoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11149.AC
25 mL, Each
Quantity
Catalog number: A11149.AC
also known as A11149-AC
Price (USD)
Price: 27.60
Online price: 23.65
Your price:
Quantity
-

Chemical Identifiers

CAS
124-07-2
IUPAC Name
octanoic acid
Molecular Formula
C8H16O2
InChI Key
WWZKQHOCKIZLMA-UHFFFAOYSA-N
SMILES
CCCCCCCC(O)=O
Appearance (Color)
Clear colorless to pale yellow
Assay (Silylated GC)
≥98.0% (UK Sourced Material)
Form
Liquid
Comment
Material Sourced in UK and US
Refractive Index
1.4260-1.4300 @ 20?C (UK Sourced Material)

Description

Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoic acid is used in the production of esters, which finds application in perfumery and also dyes manufacture. It is used as a food contact surface sanitizer in commercial food handling, as a dietary supplement and as disinfectant in health care facilities, recreational facilities, livestock premises and industrial facilities. Its acid chloride is used in the synthesis of perfluorooctanoic acid.

Solubility
Misicible with alcohol, chloroform, ether, carbon disulfide, petroleum ether and acetonitrile.Slightly miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text