2,5-Dichloroaniline, 99%
2,5-Dichloroaniline, 99%
2,5-Dichloroaniline, 99%
Thermo Scientific Chemicals

2,5-Dichloroaniline, 99%

CAS: 95-82-9 | C6H5Cl2N | 162.01 g/mol
数量:
250 g
500 g
2500 g
製品番号(カタログ番号) A11151.0E
または、製品番号A11151-0E
価格(JPY)
-
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS95-82-9
Assay (GC)≥98.5%
FormFlakes or fused solid
Appearance (Color)White to brown to grey to black
Melting Point (clear melt)46.0-52.0?C
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Sakurai, T.; Sundaralingam, M.; G. A. Jeffrey, G.A. A nuclear quadrupole resonance and X-ray study of the crystal structure of 2,5-dichloroaniline. Acta Cryst. 1963, 16, 354-363.
  2. Evgen ev, M.I.; Evgen eva, I.I.; Garmonov,S. Yu.; Ismailova, R. N. Sorption-Chromatographic Determination of Aniline, 4-Chloroaniline, and 2,5-Dichloroaniline in Air. Journal of Analytical Chemistry. 2003, 58, (6), 542-547.
  3. Nucleophilic displacement of the chloro-substituents has been carried out by heating with thiophenol in the presence of K2CO3 in NMP: J. Org. Chem., 56, 862 (1991).