Vanillin, 99%
Vanillin, 99%
Vanillin, 99%
Thermo Scientific Chemicals

Vanillin, 99%

CAS: 121-33-5 | C8H8O3 | 152.15 g/mol
製品番号(カタログ番号) A11169.36
または、製品番号A11169-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS121-33-5
IUPAC Name4-hydroxy-3-methoxybenzaldehyde
Molecular FormulaC8H8O3
InChI KeyMWOOGOJBHIARFG-UHFFFAOYSA-N
SMILESCOC1=CC(C=O)=CC=C1O
さらに表示
CommentMaterial sourced in the U.S. and in other countries
FormCrystals or crystalline powder
Appearance (Color)White to cream or pale yellow
Assay (GC)≥98.5% (non-U.S. sourced material)
CommentMaterial sourced in the U.S. and in other countries
さらに表示
Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vanillin is a flavoring agent used in sweet foods, ice cream and chocolate. It is used in perfumes as well as to control the unpleasant odors in medicines. It acts as a chemical intermediate in the preparation of pharmaceuticals. In thin layer chromatography, it is used as a general-purpose stain for developing TLC plates to enable in visualizing components of a reaction mixture. Similarly, vanillin hydro chloride staining can be used to visualize the localization of tannins in cells.

Solubility
Soluble in water, diethyl ether, acetone, methanol, benzene, petroleum ether, chloroform, carbon disulfide, glacial acetic acid, pyridine and in aqueous solutions of alkali hydroxides.

Notes
Moisture and light sensitive. Incompatible with oxidizing agents, alkalis and reducing agents.
RUO – Research Use Only

General References:

  1. Tomadoni, B.; Cassani, L.; Moreira, M. R.; Ponce, A. Efficacy of vanillin and geraniol in reducing Escherichia coli O157: H7 on strawberry juice. LWT Food Sci. Technol. 2015, 64 (2), 554-557.
  2. Di Paola, A.; Bellardita, M.; Megna, B.; Parrino, F.; Palmisano, L. Photocatalytic oxidation of trans-ferulic acid to vanillin on TiO2 and WO3-loaded TiO2 catalysts. Catal. Today 2015, 252, 195-200.