Thermo Scientific Chemicals

Methylcobalamin hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A11176.MC
100 mg, Each
Thermo Scientific Chemicals

Methylcobalamin hydrate, 97%, Thermo Scientific Chemicals

Catalog number: A11176.MC
100 mg, Each
Quantity
Catalog number: A11176.MC
also known as A11176-MC
Price (USD)
Price: 96.20
Online price: 81.65
Your price:
Quantity
-

Chemical Identifiers

CAS
288315-09-3
IUPAC Name
(10S,12R,13R,24S,35R,41R)-30,35,40-tris(2-carbamoylethyl)-24,36,41-tris(carbamoylmethyl)-46-hydroxy-12-(hydroxymethyl)-1,5,6,17,23,28,31,31,36,38,41,42-dodecamethyl-15,20-dioxo-11,14,16-trioxa-2λ⁵,9,19,26λ⁵,43,44λ⁵,45λ⁵-heptaaza-15λ⁵-phospha-1-cobaltadodecacyclo[27.14.1.1¹,³⁴.1²,⁹.1¹⁰,¹³.0¹,²⁶.0³,⁸.0²³,²⁷.0²⁵,⁴².0³²,⁴⁴.0³⁹,⁴³.0³⁷,⁴⁵]heptatetraconta-2(47),3,5,7,26,28,32(44),33,37(45),38-decaene-1,2,26,44,45-pentakis(ylium)-15-olate
Molecular Formula
C63H91CoN13O14P
InChI Key
XVLRGRFESMDSFT-BVWPOUIRNA-L
SMILES
C(CC(N)=O)[C@@H]1C(CC(N)=O)(C)C2=[N+]3[Co+]456([N+]7=CN([C@]8(O[C@@H]([C@](OP(OC(CNC(CCC9([C@H](CC(N)=O)C([N+]4=C9C(C)=C4[N+]5=C(C(C4CCC(N)=O)(C)C)C=C13)C1([C@](CC(N)=O)(C(CCC(N)=O)C(=C2C)N61)C)C)C)=O)C)([O-])=O)(C8O)[H])CO)[H])C1=CC(C)=C(C=C71)C)C
Form
Powder or crystals
Water Content (Karl Fischer Titration)
<12.0% (<10 waters of hydration)
Appearance (Color)
Red to Dark red to brown
Assay (HPLC)
≥96.0%

Description

Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcobalamin hydrate is a coenzyme required in the biosynthesis of methionine. Vitamin (hematopoietic). It acts as a histamine receptor, Alzheimer research.Methylcobalamin is also used in the treatment of peripheral neuropathy, diabetic neuropathy, and as a preliminary treatment for amyotrophic lateral sclerosis. It can be used to prevent or treat pathology arising from a lack of vitamin B12 such as pernicious anemia.

Solubility
Partially soluble in cold water, hot water.

Notes
Store at -20°C. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text