(+/-)-1,1'-Bi(2-naphthol), 99%
(+/-)-1,1'-Bi(2-naphthol), 99%
(+/-)-1,1'-Bi(2-naphthol), 99%
Thermo Scientific Chemicals

(+/-)-1,1'-Bi(2-naphthol), 99%

CAS: 602-09-5 | C20H14O2 | 286.33 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A11182.14
または、製品番号A11182-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS602-09-5
IUPAC Name[1,1'-binaphthalene]-2,2'-diol
Molecular FormulaC20H14O2
InChI KeyPPTXVXKCQZKFBN-UHFFFAOYSA-N
SMILESOC1=CC=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC=C1O
さらに表示
Appearance (Color)White to pale cream to pale brown
FormPowder
Melting Point (clear melt)214.0-221.0°C
Assay (HPLC)≥98.5%
(±)-1,1'-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1,1′-Bi(2-naphthol) acts as chiral separation agents. It is widely used in chiral liquid crystal,medicines,spices etc,.

Solubility
Soluble in dioxane (50 mg/mL). Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Refrigerated (2-8°C)
RUO – Research Use Only

General References:

  1. Lilia Kurz1; Gary Lee; David Morgans Jr; Michael Joe Waldyk;, Timothy Ward. Stereospecific functionalization of (R)-(-)-1,1'-bi-2-naphthol triflate. Tetrahedron Letters. 1990, 31 (5), 6321-6324.