Thermo Scientific Chemicals

Ammonium iron(III) citrate, brown, Thermo Scientific Chemicals

Catalog number: A11199.0B
1000 g, Each
Thermo Scientific Chemicals

Ammonium iron(III) citrate, brown, Thermo Scientific Chemicals

Catalog number: A11199.0B
1000 g, Each
Quantity
Catalog number: A11199.0B
also known as A11199-0B
Price (USD)
Price: 80.90
Online price: 68.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1185-57-5
IUPAC Name
iron(3+) 2-hydroxypropane-1,2,3-tricarboxylate amine
Molecular Formula
C6H8FeNO7
InChI Key
FRHBOQMZUOWXQL-UHFFFAOYSA-K
SMILES
N.[Fe+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Appearance (Color)
Brown-yellow to brown
Trace Metals
Pb: ≤30ppm
Form
Crystals or crystalline powder
Assay (unspecified)
Fe: 16.5-18.5%

Description

Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium iron(III) citrate, brown is used in medicine, in making blueprints, and as a feed additive. It is also used as an iron supplement and food enhancer.

Solubility
Soluble in water.1200 g/L (20°C)

Notes
Hygroscopic. Desiccate at room temperature. Store in the dark. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text