Methyl 2-chlorobenzoate, 98%
Methyl 2-chlorobenzoate, 98%
Methyl 2-chlorobenzoate, 98%
Methyl 2-chlorobenzoate, 98%
Thermo Scientific Chemicals

Methyl 2-chlorobenzoate, 98%

CAS: 610-96-8 | C8H7ClO2 | 170.59 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) A11237.36
または、製品番号A11237-36
価格(JPY)
-
見積もりを依頼する
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS610-96-8
IUPAC Namemethyl 2-chlorobenzoate
Molecular FormulaC8H7ClO2
InChI KeyJAVRNIFMYIJXIE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=CC=C1Cl
さらに表示
Refractive Index1.5340-1.5380 @ 20°C
Assay (GC)≥97.5%
FormLiquid
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only