Thermo Scientific Chemicals

Mesitylene, 98+%, Thermo Scientific Chemicals

Catalog number: A11323.AP
500 mL, Each
Thermo Scientific Chemicals

Mesitylene, 98+%, Thermo Scientific Chemicals

Catalog number: A11323.AP
500 mL, Each
Quantity
Catalog number: A11323.AP
also known as A11323-AP
Price (USD)
Quantity
-

Chemical Identifiers

CAS
108-67-8
IUPAC Name
1,3,5-trimethylbenzene
Molecular Formula
C9H12
InChI Key
AUHZEENZYGFFBQ-UHFFFAOYSA-N
SMILES
CC1=CC(C)=CC(C)=C1
Appearance (Color)
Clear colorless to very pale yellow
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.4970-1.5010 @ 20?C

Description

Mesitylene is used to make plastics and dyes. It acts as a solvent, ligand in organometallic chemistry and precursor to 2,4,6-trimethylaniline. It is also used as a developer for photopatternable silicones due to its solvent properties in the electronics industry. It is used as an internal standard in nuclear magnetic resonance (NMR) samples due to the presence of three equivalent protons in it. It is involved in the production of trimesic acid and antioxygen, epoxy firming agent and polyester resin stabilizers. Further, it is used as an additive and component in aviation gasoline blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mesitylene is used to make plastics and dyes. It acts as a solvent, ligand in organometallic chemistry and precursor to 2,4,6-trimethylaniline. It is also used as a developer for photopatternable silicones due to its solvent properties in the electronics industry. It is used as an internal standard in nuclear magnetic resonance (NMR) samples due to the presence of three equivalent protons in it. It is involved in the production of trimesic acid and antioxygen, epoxy firming agent and polyester resin stabilizers. Further, it is used as an additive and component in aviation gasoline blends.

Solubility
Partly miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text