Thermo Scientific Chemicals

p-Tolualdehyde, 98%, Thermo Scientific Chemicals

Catalog number: A11384.06
5 g, Each
Thermo Scientific Chemicals

p-Tolualdehyde, 98%, Thermo Scientific Chemicals

Catalog number: A11384.06
5 g, Each
Quantity
Catalog number: A11384.06
also known as A11384-06
Price (USD)
Price: 33.20
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
104-87-0
IUPAC Name
4-methylbenzaldehyde
Molecular Formula
C8H8O
InChI Key
FXLOVSHXALFLKQ-UHFFFAOYSA-N
SMILES
CC1=CC=C(C=O)C=C1
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Free acid (titration)
≤1.5%
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Tolualdehyde is used as an intermediate for the synthesis of pharmaceuticals, dyes perfumes and agrochemicals. It is also used as a fixative of flavorings. It is also used as an important organic intermediates, used for spices, triphenylmethane dye synthesis, etc.

Solubility
It is slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. Incompatible to Strong bases, Strong oxidizing agents, Strong reducing agents
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text