Diethyl adipate, 99%
Diethyl adipate, 99%
Diethyl adipate, 99%
Thermo Scientific Chemicals

Diethyl adipate, 99%

CAS: 141-28-6 | C10H18O4 | 202.25 g/mol
製品番号(カタログ番号) A11476.36
または、製品番号A11476-36
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS141-28-6
IUPAC Name1,6-diethyl hexanedioate
Molecular FormulaC10H18O4
InChI KeyVIZORQUEIQEFRT-UHFFFAOYSA-N
SMILESCCOC(=O)CCCCC(=O)OCC
さらに表示
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Refractive Index1.4265-1.4295 @ 20?C
FormLiquid
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Poly(vinylidene fluoride) (PVF2) gels in diethyl adipate (DEA) with fibrillar morphology is presented. Gelation process obeys the three-dimensional percolation mechanism. Combustion and emission characteristics of a direct-injection diesel engine is fueled with diesel-diethyl adipate blends.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Asok K. Dikshit,; Arun K. Nandi.Thermoreversible Gelation of Poly(vinylidene fluoride) in Diethyl Adipate: A Concerted Mechanism . Macromolecules,. 1998, 38 (25), 8886-8892.
  2. Fumio Toda,; Takaaki Suzuki,; Satoru Higa. Solvent-free Dieckmann condensation reactions of diethyl adipate and pimelate . J. Chem. Soc., Perkin Trans. 1. 1998, (21), 3521-3522.