Thermo Scientific Chemicals

Iminodiacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11510.36
500 g, Each
Thermo Scientific Chemicals

Iminodiacetic acid, 98+%, Thermo Scientific Chemicals

Catalog number: A11510.36
500 g, Each
Quantity
Catalog number: A11510.36
also known as A11510-36
Price (USD)
Price: 50.00
Online price: 41.65
Your price:
Quantity
-

Chemical Identifiers

CAS
142-73-4
IUPAC Name
2-[(carboxymethyl)amino]acetic acid
Molecular Formula
C4H7NO4
InChI Key
NBZBKCUXIYYUSX-UHFFFAOYSA-N
SMILES
OC(=O)CNCC(O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White or cream to pale brown
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%

Description

Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iminoacetic acid is used as an inhibitor of bovine liver glutamate dehydrogenase. It is used as a reagent in the synthesis of poly(N-isopropylacrylamide) (PNIPAM) microgel particles for metal affinity binding of peptides. It is mainly used for modulating peptide mobility by capillary electrophoresis. It is also used in chemotherapeutic treatment for metal poisoning. It acts as a tridendate ligand and forms metal complexes.

Solubility
Soluble in water. Insoluble in ethanol.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text