Thiazolidine-2-carboxylic acid, 97%
Thiazolidine-2-carboxylic acid, 97%
Thiazolidine-2-carboxylic acid, 97%
Thermo Scientific Chemicals

Thiazolidine-2-carboxylic acid, 97%

CAS: 16310-13-7 | C4H7NO2S | 133.165 g/mol
製品番号(カタログ番号) A11564.06
または、製品番号A11564-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS16310-13-7
IUPAC Name(2S)-1,3-thiazolidin-3-ium-2-carboxylate
Molecular FormulaC4H7NO2S
InChI KeyULSZVNJBVJWEJE-VKHMYHEASA-N
SMILES[O-]C(=O)[C@H]1[NH2+]CCS1
さらに表示
FormCrystals or powder or crystalline powder or lumps
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (Silylated GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only