Oxamide, 98+%
Oxamide, 98+%
Oxamide, 98+%
Thermo Scientific Chemicals

Oxamide, 98+%

CAS: 471-46-5 | C2H4N2O2 | 88.066 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A11570.0E
価格(JPY)
-
見積もりを依頼する
数量:
2500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS471-46-5
IUPAC Nameethanediamide
Molecular FormulaC2H4N2O2
InChI KeyYIKSCQDJHCMVMK-UHFFFAOYSA-N
SMILESNC(=O)C(N)=O
さらに表示
Appearance (Color)White
FormPowder
Assay (HPLC)≥97.5%
Elemental AnalysisCarbon C: 26.59-27.97% (Theory 27.28%)
Elemental AnalysisNitrogen N: 31.01-32.62% (Theory 31.81%)
さらに表示
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxamide is used as a stabilizer for nitrocellulose preparations. It also finds use in APCP rocket motors as a high performance burn rate suppressant.

Solubility
Soluble in ethanol, slightly soluble in water and insoluble in diethyl ether.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

General References:

  1. Yanga, L.; Liub, L.; Olsena, B.A.; Nussbaum, M.A. The determination of oxalic acid, oxamic acid, and oxamide in a drug substance by ion-exclusion chromatography. Journal of Pharmaceutical and Biomedical Analysis. 2000, 22, (3), 487-493.
  2. Isabel Fernandez.; JoseR. Pedro.; Antonio L. Rosello.; Rafael Ruiz.; Xavier Ottenwaelder.; Yves Journaux. Aerobic epoxidation of olefins catalyzed by square-planar nickel(II) complexes of bis-N,N'-disubstituted oxamides and related ligands. Tetrahedron Letters. 1998, 39, (18), 2869-2872.