4-Methylanisole, 99%
4-Methylanisole, 99%
4-Methylanisole, 99%
Thermo Scientific Chemicals

4-Methylanisole, 99%

CAS: 104-93-8 | C8H10O | 122.167 g/mol
数量:
100 mL
500 mL
2500 mL
製品番号(カタログ番号) A11572.AP
または、製品番号A11572-AP
価格(JPY)
-
数量:
500 mL
一括またはカスタム形式をリクエストする
化学物質識別子
CAS104-93-8
IUPAC Name1-methoxy-4-methylbenzene
Molecular FormulaC8H10O
InChI KeyCHLICZRVGGXEOD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C)C=C1
さらに表示
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5100-1.5130 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only