Fluorescein disodium salt hydrate
Fluorescein disodium salt hydrate
Fluorescein disodium salt hydrate
Thermo Scientific Chemicals

Fluorescein disodium salt hydrate

CAS: 518-47-8 | C20H10Na2O5 | 376.28 g/mol
数量:
100 g
500 g
2500 g
製品番号(カタログ番号) A11659.22
または、製品番号A11659-22
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS518-47-8
IUPAC Namedisodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H10Na2O5
InChI KeyNJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
さらに表示
Loss on Drying≤10.0% (105°C/constant wt., non-U.S. specification)
Loss on Drying≤10.0% (U.S. specification)
lambda-max (UV-Vis)487-505 nm (0.1 w/v% in distilled water, non-U.S. specification)
Appearance (Color)Dark red to brown or dark orange
FormPowder
さらに表示
Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.

Solubility
Soluble in water(500g/L).

Notes
Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Water-soluble form of fluorescein.