Bis(trimethylsilyl)carbodiimide, 97%
Bis(trimethylsilyl)carbodiimide, 97%
Bis(trimethylsilyl)carbodiimide, 97%
Thermo Scientific Chemicals

Bis(trimethylsilyl)carbodiimide, 97%

CAS: 1000-70-0 | C7H18N2Si2 | 186.405 g/mol
製品番号(カタログ番号) A11670.14
または、製品番号A11670-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1000-70-0
IUPAC Name2,2,6,6-tetramethyl-3,5-diaza-2,6-disilahepta-3,4-diene
Molecular FormulaC7H18N2Si2
InChI KeyKSVMTHKYDGMXFJ-UHFFFAOYSA-N
SMILESC[Si](C)(C)N=C=N[Si](C)(C)C
さらに表示
Assay (GC)≥96.0%
Refractive Index1.4320-1.4360 @ 20?C
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only