2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
2-Chloro-2-methylbutane, 95%
Thermo Scientific Chemicals

2-Chloro-2-methylbutane, 95%

CAS: 594-36-5 | C5H11Cl | 106.593 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A11703.06
または、製品番号A11703-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS594-36-5
IUPAC Name2-chloro-2-methylbutane
Molecular FormulaC5H11Cl
InChI KeyCRNIHJHMEQZAAS-UHFFFAOYSA-N
SMILESCCC(C)(C)Cl
さらに表示
Refractive Index1.4015-1.4075 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)≥94.0%
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-2-methylbutane is used as a solvent for the synthesis of other compounds and pentane. It is used in organic synthesis, Catalytic agent, Petrochemical additive.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Avoid extremes of temperature and direct sunlight.
RUO – Research Use Only
Lídia M.P.C. Albuquerque; M.Luisa C.J. Moita; M.N. Simoes; Raquel M.C. Goncalves. The enthalpies of solution of 2-chloro-2-methylpropane, 2-chloro-2-methylbutane and 3-chloro-3-methylpentane in mono- and dialcohols at 298.15 K. Thermochimica acta. 1996, 275 67-73.