Hydantoin-5-acetic acid, 98%
Hydantoin-5-acetic acid, 98%
Hydantoin-5-acetic acid, 98%
Thermo Scientific Chemicals

Hydantoin-5-acetic acid, 98%

CAS: 5427-26-9 | C5H6N2O4 | 158.11 g/mol
製品番号(カタログ番号) A11731.14
または、製品番号A11731-14
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5427-26-9
IUPAC Name2-(2,5-dioxoimidazolidin-4-yl)acetic acid
Molecular FormulaC5H6N2O4
InChI KeyDQQLZADYSWBCOX-UHFFFAOYNA-N
SMILESOC(=O)CC1NC(=O)NC1=O
さらに表示
FormCrystalline powder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Melting Point (clear melt)ca. 210-225?C dec.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only