Lithium L-lactate, 97%
Lithium L-lactate, 97%
Lithium L-lactate, 97%
Thermo Scientific Chemicals

Lithium L-lactate, 97%

CAS: 27848-80-2 | C3H5LiO3 | 96.01 g/mol
数量:
50 g
250 g
1000 g
製品番号(カタログ番号) A11818.18
または、製品番号A11818-18
価格(JPY)
-
数量:
50 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS27848-80-2
IUPAC Namelithium(1+) 2-hydroxypropanoate
Molecular FormulaC3H5LiO3
InChI KeyGKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES[Li+].CC(O)C([O-])=O
さらに表示
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Optical Rotation-14.8? ? 1? (c=1 in water)
Melting Point (clear melt)251-259?C
Appearance (Color)White
さらに表示
Useful chiral synthon, building block for depsipeptides

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only