2-Methylglutaric acid, 98%
2-Methylglutaric acid, 98%
2-Methylglutaric acid, 98%
Thermo Scientific Chemicals

2-Methylglutaric acid, 98%

CAS: 617-62-9 | C6H10O4 | 146.142 g/mol
数量:
5 g
25 g
100 g
製品番号(カタログ番号) A11823.06
または、製品番号A11823-06
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS617-62-9
IUPAC Name2-methylpentanedioic acid
Molecular FormulaC6H10O4
InChI KeyAQYCMVICBNBXNA-UHFFFAOYNA-N
SMILESCC(CCC(O)=O)C(O)=O
さらに表示
Appearance (Color)White to pale cream or pale yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Melting Point (clear melt)71.0-80.0°C
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylglutaric acid is used as primary and secondary intermediates in pharmaceutical and chemical research.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Incompatible with bases and oxidizing agents.
RUO – Research Use Only
J.L. Willems; A.F.M. de Kort; T.B. Vree; J.M.F. Trijbels; J.H. Veerkamp; L.A.H. Monnens. Non-enzymic conversion of pyruvate in aqueous solution to 2,4-dihydroxy-2-methylglutaric acid. FEBS letters. 1978, 86 42-44.