Thermo Scientific Chemicals

Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A11948.22
100 g, Each
Thermo Scientific Chemicals

Cyclopropanecarbonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A11948.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A11948.22
also known as A11948-22
Price (USD)
Price: 147.00
Online price: 127.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4023-34-1
IUPAC Name
cyclopropanecarbonyl chloride
Molecular Formula
C4H5ClO
InChI Key
ZOOSILUVXHVRJE-UHFFFAOYSA-N
SMILES
ClC(=O)C1CC1
Assay (GC)
≥97.5%
Refractive Index
1.4510-1.4550 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms

Description

It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is primarily used an intermediate in the manufacture of pharmaceutical and agrochemical products.

Solubility
Soluble in water (reacts violently).

Notes
Moisture Sensitive. Store away from oxidizing agents, bases, active metals and water/moisture. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text