Thermo Scientific Chemicals

Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals

Catalog number: A12000.06
5 g, Each
Thermo Scientific Chemicals

Pentafluoroiodobenzene, 97%, Thermo Scientific Chemicals

Catalog number: A12000.06
5 g, Each
Quantity
Catalog number: A12000.06
also known as A12000-06
Price (USD)
Price: 34.40
Online price: 32.65
Your price:
Quantity
-

Chemical Identifiers

CAS
827-15-6
IUPAC Name
1,2,3,4,5-pentafluoro-6-iodobenzene
Molecular Formula
C6F5I
InChI Key
OPYHNLNYCRZOGY-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(I)C(F)=C1F
Form
Liquid
Appearance (Color)
Clear colorless to pink
Assay (GC)
≥96.0%
Refractive Index
1.4930-1.4980 @ 20°C
Identification (FTIR)
Conforms

Description

Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Iodopentafluorobenzene was used to study the formation of radical ions of iodopentafluorobenzene in aqueous solution. It was used as solvent in a study to determine singlet oxygen lifetimes from phosphorescence decays in halogen substituted perfluorinated solvents by infrared emission spectrometery. It has potential applications in plasma processing industry and in preparation of catalysts.

Solubility
Insoluble in water.

Notes
Light sensitive. Store at room temperature. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text