Thermo Scientific Chemicals

3-Chloroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A12019.36
500 g, Each
Thermo Scientific Chemicals

3-Chloroaniline, 99%, Thermo Scientific Chemicals

Catalog number: A12019.36
500 g, Each
Quantity
Catalog number: A12019.36
also known as A12019-36
Price (USD)
Quantity
-

Chemical Identifiers

CAS
108-42-9
IUPAC Name
3-chloroaniline
Molecular Formula
C6H6ClN
InChI Key
PNPCRKVUWYDDST-UHFFFAOYSA-N
SMILES
NC1=CC=CC(Cl)=C1
Appearance (Color)
Clear colorless to yellow to brown
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.5925-1.5955 @ 20?C

Description

3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloroaniline was the only aniline compound to increase plasmaALT/GPT activity at 48 h. It is employed in the bioaugmentation of activated sludge.

Solubility
Soluble in water. [6.8 g/L (20°C)].

Notes
Store in cool place. Keep away from oxidizing agents. Incompatible with bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text